Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMO1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117281.3 | NP_192948.1 | 3 - 4 | 982 | 102 | |
GenBank mRNA | AY125499.1 | - | 3 - 4 | 960 | 102 | |
BT000546.1 | - | 3 - 4 | 814 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |