Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129018.4 | NP_181011.2 | 8 - 9 | 1656 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |