Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336523.1 | NP_001324697.1 | 27 - 28 | 4259 | 104 | |
NM_129022.8 | NP_181015.7 | 27 - 28 | 4070 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |