Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129030.2 | NP_181023.1 | 2 - 3 | 288 | 98 | |
GenBank mRNA | AF021804.1 | - | 1 - 2 | 49 | 98 | |
AY050326.1 | - | 2 - 3 | 89 | 98 | ||
BT000482.1 | - | 1 - 2 | 49 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |