Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036157.2 | NP_001031234.1 | 1 - 2 | 297 | 141 | |
NM_105155.4 | NP_176661.1 | 2 - 3 | 308 | 141 | ||
GenBank mRNA | AK227053.1 | - | 2 - 3 | 276 | 141 | |
AY466161.1 | - | 2 - 3 | 269 | 141 | ||
BT028969.1 | - | 1 - 2 | 98 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |