Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334181.1 | NP_001319322.1 | 4 - 5 | 1424 | 122 | |
NM_105160.3 | NP_176666.1 | 4 - 5 | 1429 | 122 | ||
GenBank mRNA | AF015542.1 | - | 4 - 5 | 1149 | 122 | |
AK316773.1 | - | 4 - 5 | 1169 | 122 | ||
AY062526.1 | - | 4 - 5 | 1163 | 122 | ||
AY093306.1 | - | 3 - 4 | 962 | 122 | ||
D89993.1 | - | 4 - 5 | 1166 | 122 | ||
Y14252.1 | - | 1160 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |