Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110994.4 | NP_186777.1 | 3 - 4 | 425 | 87 | |
GenBank mRNA | AY037217.1 | - | 3 - 4 | 339 | 87 | |
AY113169.1 | - | 3 - 4 | 222 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |