Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110999.7 | NP_186782.2 | 9 - 10 | 695 | 114 | |
GenBank mRNA | AB074532.1 | - | 9 - 10 | 624 | 114 | |
AY093024.1 | - | 9 - 10 | 654 | 114 | ||
BT000412.1 | - | 9 - 10 | 624 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |