Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340813.1 | NP_001329616.1 | 1 - 2 | 435 | 122 | |
NM_117360.8 | NP_193027.5 | 1 - 2 | 478 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |