Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340844.1 | NP_001328364.1 | 2 - 3 | 1604 | 83 | |
NM_001340845.1 | NP_001328363.1 | 4 - 5 | 1317 | 83 | ||
NM_117399.4 | NP_193062.1 | 3 - 4 | 1502 | 83 | ||
GenBank mRNA | AY133691.1 | - | 3 - 4 | 1048 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |