Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117482.4 | NP_193143.2 | 5 - 6 | 959 | 120 | |
GenBank mRNA | AY062804.1 | - | 5 - 6 | 928 | 120 | |
BT010332.1 | - | 5 - 6 | 810 | 120 | ||
EF093797.1 | - | 5 - 6 | 810 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |