Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117499.5 | NP_193158.2 | 7 - 8 | 1226 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |