Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340918.1 | NP_001328596.1 | 4 - 5 | 677 | 109 | |
NM_117511.5 | NP_193170.1 | 4 - 5 | 677 | 109 | ||
GenBank mRNA | AF250343.1 | - | 4 - 5 | 501 | 109 | |
AY052364.1 | - | 4 - 5 | 647 | 109 | ||
AY060528.1 | - | 4 - 5 | 347 | 109 | ||
AY943850.1 | - | 4 - 5 | 347 | 109 | ||
X78818.1 | - | 518 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |