Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203809.1 | NP_001190738.2 | 3 - 4 | 527 | 98 | |
NM_117667.9 | NP_193311.6 | 3 - 4 | 570 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |