Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341093.1 | NP_001319957.1 | 4 - 5 | 3999 | 66 | |
NM_001341094.1 | NP_001328665.1 | 4 - 5 | 4008 | 66 | ||
NM_001341095.1 | NP_001328666.1 | 4 - 5 | 4008 | 66 | ||
NM_001341096.1 | NP_001328664.1 | 4 - 5 | 3951 | 66 | ||
NM_117726.5 | NP_193364.5 | 3 - 4 | 3595 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |