Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203815.1 | NP_001190744.1 | 3 - 4 | 475 | 58 | |
NM_117778.8 | NP_567513.4 | 3 - 4 | 513 | 58 | ||
GenBank mRNA | AF057044.1 | - | 3 - 4 | 384 | 58 | |
AY058849.2 | - | 3 - 4 | 441 | 58 | ||
BT001067.1 | - | 3 - 4 | 308 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |