Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341198.1 | NP_001328216.1 | 5 - 6 | 1032 | 78 | |
NM_117835.5 | NP_193462.1 | 5 - 6 | 864 | 78 | ||
GenBank mRNA | AY078967.1 | - | 5 - 6 | 841 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |