Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341257.1 | NP_001328203.1 | 9 - 10 | 2947 | 83 | |
NM_117915.7 | NP_193539.6 | 9 - 10 | 2528 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |