Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117956.3 | NP_193578.1 | 1 - 2 | 384 | 107 | |
GenBank mRNA | BT010933.1 | - | 1 - 2 | 281 | 107 | |
BT011645.1 | - | 1 - 2 | 224 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |