Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105215.3 | NP_564850.1 | 1 - 2 | 378 | 80 | |
GenBank mRNA | AF361573.1 | - | 1 - 2 | 378 | 80 | |
AY093985.1 | - | 1 - 2 | 185 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |