Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105215.3 | NP_564850.1 | 2 - 3 | 512 | 131 | |
GenBank mRNA | AF361573.1 | - | 2 - 3 | 512 | 131 | |
AY093985.1 | - | 2 - 3 | 319 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |