Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105233.5 | NP_176737.2 | 12 - 13 | 1628 | 73 | |
GenBank mRNA | AY128283.1 | - | 12 - 13 | 1202 | 73 | |
BT000831.1 | - | 12 - 13 | 1184 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |