Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334346.1 | NP_001322211.1 | 8 - 9 | 1324 | 123 | |
NM_105480.3 | NP_564921.1 | 8 - 9 | 1333 | 123 | ||
GenBank mRNA | AF216524.1 | - | 8 - 9 | 1158 | 123 | |
AK229148.1 | - | 8 - 9 | 1215 | 123 | ||
AY084533.1 | - | 8 - 9 | 1229 | 123 | ||
BT001920.1 | - | 8 - 9 | 1108 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |