Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105485.4 | NP_176982.1 | 3 - 4 | 722 | 58 | |
GenBank mRNA | AB493525.1 | - | 3 - 4 | 701 | 58 | |
AY052645.1 | - | 3 - 4 | 701 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |