Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105575.3 | NP_177067.1 | 4 - 5 | 671 | 128 | |
NM_202378.2 | NP_974107.1 | 5 - 6 | 586 | 128 | ||
GenBank mRNA | AF528060.1 | - | 4 - 5 | 332 | 128 | |
BT029177.1 | - | 4 - 5 | 298 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |