Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105649.3 | NP_564976.1 | 1 - 2 | 200 | 90 | |
GenBank mRNA | AB493530.1 | - | 1 - 2 | 61 | 90 | |
AF404859.1 | - | 1 - 2 | 61 | 90 | ||
BT029234.1 | - | 1 - 2 | 61 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |