Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105651.5 | NP_564977.1 | 9 - 10 | 2136 | 88 | |
GenBank mRNA | AY050398.1 | - | 9 - 10 | 2012 | 88 | |
BT000643.1 | - | 9 - 10 | 1966 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |