Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334433.1 | NP_001323064.1 | 15 - 16 | 3506 | 152 | |
NM_105674.6 | NP_177163.3 | 16 - 17 | 3339 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |