Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105701.2 | NP_564987.2 | 1 - 2 | 628 | 84 | |
GenBank mRNA | AY074345.1 | - | 1 - 2 | 626 | 84 | |
BT003008.1 | - | 1 - 2 | 617 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |