Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105840.4 | NP_565026.1 | 16 - 17 | 1753 | 82 | |
GenBank mRNA | AY046014.1 | - | 16 - 17 | 1674 | 82 | |
AY133819.1 | - | 15 - 16 | 1405 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |