Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 20 - 21 | 2132 | 90 | |
NM_105840.4 | NP_565026.1 | 20 - 21 | 2051 | 90 | ||
GenBank mRNA | AY046014.1 | - | 20 - 21 | 1972 | 90 | |
AY133819.1 | - | 19 - 20 | 1703 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |