Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105840.4 | NP_565026.1 | 8 - 9 | 950 | 98 | |
GenBank mRNA | AY046014.1 | - | 8 - 9 | 871 | 98 | |
AY133819.1 | - | 7 - 8 | 602 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |