Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036189.1 | NP_001031266.1 | 1 - 2 | 350 | 64 | |
NM_105844.4 | NP_177331.1 | 1 - 2 | 384 | 64 | ||
NM_202398.1 | NP_974127.1 | 1 - 2 | 332 | 64 | ||
GenBank mRNA | AY070403.1 | - | 1 - 2 | 332 | 64 | |
AY117240.1 | - | 1 - 2 | 195 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |