Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105844.4 | NP_177331.1 | 8 - 9 | 1218 | 112 | |
NM_202398.1 | NP_974127.1 | 8 - 9 | 1161 | 112 | ||
GenBank mRNA | AY070403.1 | - | 8 - 9 | 1166 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |