Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105926.5 | NP_177411.1 | 3 - 4 | 749 | 87 | |
GenBank mRNA | AK175909.1 | - | 3 - 4 | 739 | 87 | |
BT010652.1 | - | 3 - 4 | 543 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |