Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105930.3 | NP_177415.1 | 2 - 3 | 476 | 107 | |
GenBank mRNA | AY059850.1 | - | 2 - 3 | 402 | 107 | |
AY943843.1 | - | 2 - 3 | 120 | 107 | ||
BT010328.1 | - | 2 - 3 | 120 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |