Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344150.1 | NP_001329981.1 | 35 - 36 | 4010 | 120 | |
NM_001344151.1 | NP_001329982.1 | 35 - 36 | 4010 | 120 | ||
NM_001344152.1 | NP_001329979.1 | 34 - 35 | 4081 | 120 | ||
NM_001344153.1 | NP_001329980.1 | 34 - 35 | 3927 | 120 | ||
NM_123045.4 | NP_198503.3 | 34 - 35 | 3930 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |