Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123064.3 | NP_568545.1 | 3 - 4 | 440 | 73 | |
GenBank mRNA | AF378857.1 | - | 3 - 4 | 367 | 73 | |
AK175306.1 | - | 3 - 4 | 357 | 73 | ||
AY050464.1 | - | 3 - 4 | 314 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |