Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339103.1 | NP_001327387.1 | 2 - 3 | 956 | 81 | |
NM_001339104.1 | NP_001327388.1 | 2 - 3 | 946 | 81 | ||
NM_114238.3 | NP_189956.2 | 2 - 3 | 956 | 81 | ||
GenBank mRNA | BT029752.1 | - | 2 - 3 | 766 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |