Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339194.1 | NP_001327790.1 | 7 - 8 | 1257 | 109 | |
NM_001339195.1 | NP_001327789.1 | 6 - 7 | 1164 | 109 | ||
NM_001339196.1 | NP_001327788.1 | 6 - 7 | 1364 | 109 | ||
NM_001339197.1 | NP_001327787.1 | 7 - 8 | 1257 | 109 | ||
NM_114382.5 | NP_190099.3 | 6 - 7 | 1145 | 109 | ||
GenBank mRNA | AB247155.1 | - | 6 - 7 | 1105 | 109 | |
DQ508794.2 | - | 6 - 7 | 1105 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |