Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335236.1 | NP_001324657.1 | 2 - 3 | 570 | 108 | |
NM_126476.3 | NP_178524.2 | 1 - 2 | 248 | 108 | ||
GenBank mRNA | BT010760.1 | - | 1 - 2 | 248 | 108 | |
BT011326.1 | - | 1 - 2 | 205 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |