Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 26 - 27 | 3121 | 120 | |
NM_001335346.1 | NP_001323940.1 | 26 - 27 | 3121 | 120 | ||
NM_001335347.1 | NP_001323936.1 | 25 - 26 | 3038 | 120 | ||
NM_001335348.1 | NP_001323937.1 | 25 - 26 | 2967 | 120 | ||
NM_001335349.1 | NP_001323938.1 | 23 - 24 | 2898 | 120 | ||
NM_126770.8 | NP_178811.7 | 25 - 26 | 2978 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |