Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124823.2 | NP_001118295.1 | 15 - 16 | 2109 | 99 | |
NM_001335364.1 | NP_001324918.1 | 15 - 16 | 2084 | 99 | ||
NM_126831.4 | NP_178872.2 | 15 - 16 | 2166 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |