Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036274.1 | NP_001031351.1 | 7 - 8 | 1023 | 69 | |
NM_126893.4 | NP_178937.2 | 7 - 8 | 1023 | 69 | ||
GenBank mRNA | BT004306.1 | - | 7 - 8 | 1023 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |