Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335389.1 | NP_001325087.1 | 30 - 31 | 5643 | 157 | |
NM_001335390.1 | NP_001325086.1 | 30 - 31 | 5039 | 157 | ||
NM_126926.5 | NP_178970.3 | 30 - 31 | 5237 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |