Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084418.1 | NP_001077887.1 | 11 - 12 | 1115 | 64 | |
NM_001084419.1 | NP_001077888.1 | 12 - 13 | 1315 | 64 | ||
NM_126989.2 | NP_179032.1 | 12 - 13 | 1315 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |