Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMO2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100616.3 | NP_563789.1 | 6 - 7 | 1082 | 78 | |
NM_202048.2 | NP_973777.1 | 5 - 6 | 1054 | 78 | ||
GenBank mRNA | AF346734.1 | - | 6 - 7 | 727 | 78 | |
AK316820.1 | - | 5 - 6 | 1055 | 78 | ||
BT044610.1 | - | 6 - 7 | 727 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |