Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035914.2 | NP_001030991.2 | 6 - 7 | 740 | 113 | |
NM_001084012.1 | NP_001077481.1 | 5 - 6 | 732 | 113 | ||
NM_001084013.1 | NP_001077482.1 | 6 - 7 | 648 | 113 | ||
NM_001123772.2 | NP_001117244.1 | 5 - 6 | 808 | 113 | ||
NM_001331739.1 | NP_001318949.1 | 5 - 6 | 896 | 113 | ||
NM_100663.4 | NP_172267.1 | 5 - 6 | 781 | 113 | ||
NM_179276.2 | NP_849607.1 | 6 - 7 | 778 | 113 | ||
NM_202057.2 | NP_973786.1 | 5 - 6 | 533 | 113 | ||
GenBank mRNA | AK230096.1 | - | 1186 | 113 | ||
AY039879.1 | - | 5 - 6 | 584 | 113 | ||
AY056395.1 | - | 5 - 6 | 584 | 113 | ||
AY086425.1 | - | 5 - 6 | 637 | 113 | ||
AY094002.1 | - | 4 - 5 | 494 | 113 | ||
X59600.1 | - | 533 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |