Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102230.5 | NP_173794.3 | 8 - 9 | 938 | 81 | |
NM_202171.2 | NP_973900.2 | 8 - 9 | 1087 | 81 | ||
GenBank mRNA | AJ251296.1 | - | 8 - 9 | 995 | 81 | |
AY062593.1 | - | 8 - 9 | 1006 | 81 | ||
AY093360.1 | - | 8 - 9 | 938 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |