Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036016.2 | NP_001031093.1 | 4 - 5 | 686 | 134 | |
NM_001332707.1 | NP_001323071.1 | 4 - 5 | 923 | 134 | ||
NM_001332708.1 | NP_001323072.1 | 4 - 5 | 1122 | 134 | ||
NM_001332709.1 | NP_001323073.1 | 4 - 5 | 1082 | 134 | ||
NM_102390.3 | NP_173950.1 | 3 - 4 | 1189 | 134 | ||
NM_202184.3 | NP_973913.1 | 4 - 5 | 933 | 134 | ||
GenBank mRNA | AJ630483.1 | - | 4 - 5 | 626 | 134 | |
AK117355.1 | - | 3 - 4 | 889 | 134 | ||
AY568655.1 | - | 4 - 5 | 626 | 134 | ||
BT005334.1 | - | 3 - 4 | 779 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |